MilliporeSigma
All Photos(1)

F5886

Supelco

Fentanyl citrate salt solution

drug standard, 100 μg/mL in methanol: tert-butanol (3:2)

CAS Number:
MDL number:
PubChem Substance ID:

grade

drug standard

assay

≥98%

drug control

Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

concentration

100 μg/mL in methanol: tert-butanol (3:2)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCC(=O)N(C1CCN(CC1)CCc2ccccc2)c3ccccc3

InChI

1S/C22H28N2O.C6H8O7/c1-2-22(25)24(20-11-7-4-8-12-20)21-14-17-23(18-15-21)16-13-19-9-5-3-6-10-19;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-12,21H,2,13-18H2,1H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

IVLVTNPOHDFFCJ-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Teresa J Carrette et al.
Diving and hyperbaric medicine, 43(1), 9-15 (2013-03-20)
We reviewed the occurrence, trends, definition and severity of the Irukandji syndrome for the Cairns region of North Queensland, Australia. A retrospective analysis of patient files from two sources was conducted: historic accounts kept by Dr Jack Barnes for the
John E Poulos et al.
AANA journal, 81(1), 31-36 (2013-03-22)
This retrospective cohort study evaluated procedural efficiency and patient satisfaction in patients who had received propofol, midazolam/fentanyl/propofol (MFP), or midazolam/fentanyl, as sedation for either esophagogastroduodenoscopy or colonoscopy. Questionnaires about procedural times and patient satisfaction were administered. Use of propofol for
An EMS guide to chronic pain.
Kevin T Collopy et al.
EMS world, 42(7), 59-63 (2013-08-10)
Hong-Xia Zhu et al.
Zhongguo zhen jiu = Chinese acupuncture & moxibustion, 33(4), 342-346 (2013-07-04)
To observe the clinical effect of acupoint injection with small dose of fentanyl-droperido mixed liquor at different time stages on labor analgesia, and explore its function mechanism. One hundred and fifty cases of full-term primiparas who were intended to take
[Pain therapy. Treating pain patients with opioid preparations].
MMW Fortschritte der Medizin, 155(6), 70-71 (2013-05-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service