All Photos(1)

G6262

Sigma-Aldrich

Glycolithocholic acid 3-sulfate disodium salt

Synonym(s):
3α-Hydroxy-5β-cholan-24-oic acid N-(carboxymethyl)amide 3-sulfate, Sulfoglycolithocholic acid
Linear Formula:
C26H41NO7SNa2
CAS Number:
Molecular Weight:
557.65
MDL number:

description

anionic

SMILES string

[Na].C[C@H](CCC(=O)NCC(O)=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@@H](CC[C@]4(C)[C@H]3CC[C@]12C)OS(O)(=O)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service