MilliporeSigma
All Photos(3)

G6922

Sigma-Aldrich

Anti-GOT1 (57-70) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):
Anti-GIG18, Anti-GOT1 Aspartate aminotransferase, cytoplasmic

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~46 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GOT1(2805)

Immunogen

synthetic peptide corresponding to amino acids 57-70 of human GOT1

Application

Anti-GOT1 antibody produced in rabbit is suitable for western blot analysis at a working dilution of 1:500-1:2000.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

GOT1 (glutamic-oxaloacetic transaminase 1) gene encodes an enzyme that is pyridoxal phosphate-dependent. It is localized to the cytoplasm. It has an essential role in amino acid metabolism, urea and tricarboxylic acid cycles. It is involved in the biosynthesis of L-glutamate from L-aspartate or L-cysteine. Glutamate oxaloacetate transaminase can be used to metabolize excessive amounts of glutamate in blood during ischemic stroke.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service