G7788

Sigma-Aldrich

α-Guanidinoglutaric acid

≥98%

Synonym(s):
GGA
Empirical Formula (Hill Notation):
C6H11N3O4
CAS Number:
Molecular Weight:
189.17
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥98%

storage temp.

2-8°C

SMILES string

NC(=N)NC(CCC(O)=O)C(O)=O

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Daiki D Horikawa et al.
PloS one, 8(6), e64793-e64793 (2013-06-14)
Tardigrades inhabiting terrestrial environments exhibit extraordinary resistance to ionizing radiation and UV radiation although little is known about the mechanisms underlying the resistance. We found that the terrestrial tardigrade Ramazzottius varieornatus is able to tolerate massive doses of UVC irradiation...
Haihui Liao et al.
PloS one, 6(12), e28582-e28582 (2011-12-17)
Meesmann epithelial corneal dystrophy (MECD) is an inherited eye disorder caused by dominant-negative mutations in either keratins K3 or K12, leading to mechanical fragility of the anterior corneal epithelium, the outermost covering of the eye. Typically, patients suffer from lifelong...
Ahmad Amanzada et al.
PloS one, 8(12), e84026-e84026 (2014-01-01)
Eradication of hepatitis C virus (HCV) infection, both spontaneous and treatment-induced, is marked by the wildtype allele C of a single nucleotide polymorphism upstream of the IL28B gene, rs12979860. This favorable allele was recently described to be in linkage disequilibrium...
Guanxiong Ding et al.
PloS one, 8(10), e77400-e77400 (2013-10-17)
Though prostate cancer (PCa) has slow progression, the hormone refractory (HRCP) and metastatic entities are substantially lethal and lack effective treatments. Transcription factor Slug is critical in regulating metastases of various tumors including PCa. Here we studied targeted therapy against...
Philippa K Harris et al.
PLoS pathogens, 1(3), 241-251 (2005-12-03)
Proteolytic shedding of surface proteins during invasion by apicomplexan parasites is a widespread phenomenon, thought to represent a mechanism by which the parasites disengage adhesin-receptor complexes in order to gain entry into their host cell. Erythrocyte invasion by merozoites of...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.