G8628

Sigma-Aldrich

18β-Glycyrrhetinic acid

~95%

Synonym(s):
3β-Hydroxy-11-oxo-18β,20β-olean-12-en-29-oic acid
EC Number:

assay

~95%

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service