GW21475B

Sigma-Aldrich

Anti-PTPRJ (ab2) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):
Anti-Receptor-type tyrosine-protein phosphatase η, Anti-CD148

biological source

chicken

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21475B

application(s)

western blot: suitable

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PTPRJ(5795)

Immunogen

Recombinant protein tyrosine phosphatase, receptor type, J
Immunogen Sequence: GI # 18860900, sequence 997-1337

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service