Documents

H133

Sigma-Aldrich

N,N,N-Trimethylserotonin iodide

solid

Synonym(s):
5-HTQ Iodide, Trimethylserotonin iodide
Empirical Formula (Hill Notation):
C13H19IN2O
Molecular Weight:
346.21
MDL number:
Pricing and availability is not currently available.

form

solid

color

tan

solubility

H2O: soluble

storage temp.

2-8°C

Biochem/physiol Actions

5-HT3 serotonin receptor agonist that does not cross the blood-brain barrier; quaternary analog of serotonin.

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.