All Photos(1)

H1625

Sigma-Aldrich

Hematein

from logwood, ~85%

Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
Colour Index Number:
75290
Beilstein:
93039
EC Number:
MDL number:

biological source

logwood

assay

~85%

storage temp.

2-8°C

SMILES string

Oc1ccc2c(OCC3(O)CC4=CC(=O)C(O)=CC4=C23)c1O

InChI

1S/C16H12O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,17,19-21H,5-6H2

InChI key

HLUCICHZHWJHLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Sometimes called hematine or haematein; do not confuse with hematin, which is a derivative of hemin.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service