H3883

Sigma-Aldrich

Hemoglobin from rat

lyophilized powder

Synonym(s):
Hb
CAS Number:
MDL number:
Pricing and availability is not currently available.

biological source

rat

form

lyophilized powder

storage temp.

2-8°C

Gene Information

rat ... Hba-a1(25632), Hbb(24440), Hbz(287168)

Biochem/physiol Actions

Oxygen transporter, NO scavenger

Caution

Since native hemoglobin is readily oxidized in air, these preparations may be predominantly methemoglobin.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Tianzhu Zang et al.
Analytical chemistry, 81(21), 9065-9071 (2009-10-31)
Elevated blood levels of homocysteine (Hcy), hyperhomocysteinemia or homocystinuria, have been associated with various diseases and conditions. Homocysteine thiolactone (Hcy TL) is a metabolite of Hcy and reacts with amine groups in proteins to form stable amides, homocystamides, or N-homocysteinylated...
Vladimir M Turzhitsky et al.
Applied optics, 47(32), 6046-6057 (2008-11-13)
There has been significant interest in developing depth-selective optical interrogation of biological tissue in general and of superficial (e.g., mucosal) tissue in particular. We report an in vivo polarization-gating fiber-optic probe that obtains backscattering spectroscopic measurements from a range of...
Hiroshi Hori et al.
Journal of inorganic biochemistry, 116, 53-54 (2012-09-27)
The effect of the quaternary structure change in deoxy-Hb on the electronic states of the ferrous high-spin heme was studied by multi-frequency electron paramagnetic resonance (MFEPR). We used a single-pass transmission-type EPR at 70-220 GHz and at 1.5K. We compared...
Rebekka Heitmar et al.
Investigative ophthalmology & visual science, 54(4), 2807-2811 (2013-03-16)
To establish the optimal flash settings for retinal vessel oxygen saturation parameters using dual-wavelength imaging in a multiethnic group. Twelve healthy young subjects (mean age 32 years [SD 7]; three Mediterranean, two South Asian, and seven Caucasian individuals) underwent retinal...
Bensachee Pattamadilok et al.
Photodermatology, photoimmunology & photomedicine, 28(4), 213-215 (2012-09-29)
Tristimulus colorimetry and diffuse reflectance spectroscopy (DRS) are white-light skin reflectance techniques used to measure the intensity of skin pigmentation. The tristimulus colorimeter is an instrument that measures a perceived color and the DRS instrument measures biological chromophores of the...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.