All Photos(1)

H6014

Supelco

Methyl hentriacontanoate

~98%

Synonym(s):
Hentriacontanoic acid methyl ester
Linear Formula:
CH3(CH2)29COOCH3
CAS Number:
Molecular Weight:
480.85
Beilstein:
5078115
MDL number:

assay

~98%

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service