All Photos(1)

H6877

Sigma-Aldrich

12-Hydroxyoctadecanoic acid

≥99%

Synonym(s):
12-Hydroxystearic acid, 12-HSA
Linear Formula:
CH3(CH2)5CH(OH)(CH2)10COOH
CAS Number:
Molecular Weight:
300.48
Beilstein:
1726730
EC Number:
MDL number:

assay

≥99%

mp

74-76 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCCC(O)CCCCCCCCCCC(O)=O

InChI

1S/C18H36O3/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21/h17,19H,2-16H2,1H3,(H,20,21)

InChI key

ULQISTXYYBZJSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service