H7255

Sigma-Aldrich

Hemoglobin from rabbit

lyophilized powder

CAS Number:
MDL number:
Pricing and availability is not currently available.

biological source

rabbit

form

lyophilized powder

storage temp.

2-8°C

Gene Information

rabbit ... HBA(100009249), HBB2(100009084)

Biochem/physiol Actions

Oxygen transporter, NO scavenger

Caution

Since native hemoglobin is readily oxidized in air, these preparations may be predominantly methemoglobin.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Vladimir M Turzhitsky et al.
Applied optics, 47(32), 6046-6057 (2008-11-13)
There has been significant interest in developing depth-selective optical interrogation of biological tissue in general and of superficial (e.g., mucosal) tissue in particular. We report an in vivo polarization-gating fiber-optic probe that obtains backscattering spectroscopic measurements from a range of...
Omar Falou et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2012, 3155-3158 (2013-02-01)
The necessity for a non-invasive and inexpensive imaging modality to both diagnose and monitor treatment response has lead to renewed interest in the potential of optical imaging. The aim of this study was to investigate the potential of diffuse optical...
Luca Pollonini et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2012, 3760-3763 (2013-02-01)
Current methods for monitoring exercise exertion rely upon heart rate monitors, which represent a crude and lagging indicator of conditioning. The rationale for the present study is that both systemic and local metabolic mechanisms are responsible for physical performance, and...
J L Schei et al.
Neuroscience, 227, 191-200 (2012-10-09)
Coupling between neural activity and hemodynamic responses is important in understanding brain function, interpreting brain-imaging signals, and assessing pathological conditions. Tissue state is a major factor in neurovascular coupling and may alter the relationship between neural and hemodynamic activity. However...
R Bokiniec et al.
Advances in medical sciences, 57(2), 348-355 (2012-11-20)
The aim of this study was to determine brain oxygenation in full-term and preterm neonates using near infrared spectroscopy. A total of 88 full-term and preterm newborn infants without hypoxic-ischaemic disorders admitted to the NICU were examined using NIRS on...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.