Documents

H9528

Sigma-Aldrich

(±)-(E)-2-(E)-Hydroxyimino-6-methoxy-4-methyl-5-nitro-3-hexenamide

≥95% (HPLC)

Synonym(s):
NOR-1
Empirical Formula (Hill Notation):
C8H13N3O5
CAS Number:
Molecular Weight:
231.21
MDL number:
Pricing and availability is not currently available.

assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

COCC(\C(C)=C\C(=N/O)C(N)=O)N+(O-)=O

Application

NO donor; releases nitric oxide spontaneously in a rate-controlled manner; half life in solution = 1.7 min.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xi

Risk Statement

36/37/38

Safety Statement

26-36

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Y Kita et al.
European journal of pharmacology, 257(1-2), 123-130 (1994-05-12)
(+-)-(E)-Ethyl-2-[(E)-hydroxyimino]-5-nitro-3-hexeneamide (FK409), which was isolated from microbial products, has been reported to show a vasorelaxant effect through a mechanism similar to that of the organic nitrates such as isosorbide dinitrate. In solution at pH 7.4, FK409 decomposed and released nitric...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.