HPA013653

Sigma-Aldrich

Anti-LRFN2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):
Anti-Leucine-rich repeat and fibronectin type-III domain-containing protein 2 precursor, Anti-Synaptic adhesion-like molecule 1
Human Protein Atlas Number:
Pricing and availability is not currently available.

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

application(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

LPHLSNSTSRTAPPKSRLSDITGSSKTSRGGGGSGGGEPPKSPPERAVLVSEVTTTSALVKWSVSKSAPRVKMYQLQYNCSDDEVLIYRMIPASNKAFVVNNLVSGTGYDLCVLA

conjugate

unconjugated

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LRFN2(57497)

Immunogen

Leucine-rich repeat and fibronectin type-III domain-containing protein 2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project (www.proteinatlas.org)and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

The gene LRFN2 (leucine rich repeat and fibronectin type III domain containing 2) encodes a neuronal transmembrane protein that is expressed in the bone marrow. It may function in normal hematopoiesis and the regulation of colony forming units-fibroblast. Along with the protein Myc, it facilitates erythroblastosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72080.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Sigma-Aldrich Co. LLC

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

1

Certificate of Analysis
Certificate of Origin
Andres Castellanos et al.
Experimental hematology, 35(5), 724-734 (2007-06-20)
The transgenic mouse line MEnTCD2.5 expresses a dominant interfering Myb protein in a T-cell-specific fashion. When MEnTCD2.5 animals are crossed to a second line ubiquitously expressing Myc, they develop a rapid onset, fatal disease characterized by enlarged lymph nodes full...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.