MilliporeSigma
  • I1397
All Photos(1)

I1397

Sigma-Aldrich

Barium ionophore I

Synonym(s):
N,N,N′,N′-Tetracyclohexyl-oxybis(o-phenyleneoxy)diacetamide, V 163
Empirical Formula (Hill Notation):
C40H56N2O5
CAS Number:
Molecular Weight:
644.88
Beilstein:
6554767
MDL number:

storage temp.

2-8°C

SMILES string

O=C(COc1ccccc1Oc2ccccc2OCC(=O)N(C3CCCCC3)C4CCCCC4)N(C5CCCCC5)C6CCCCC6

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service