All Photos(1)

I6139

Sigma-Aldrich

Anti-IKKα antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):
Anti-IκB Kinase α
MDL number:

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 85 kDa

species reactivity

human

application(s)

microarray: suitable
western blot: 1:1,000 using whole extract of human acute lymphoma Jurkat cells and human breast adenocarcinoma MCF7 cells

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CHUK(1147)
mouse ... Chuk(12675)

Immunogen

synthetic peptide located near the C-terminal region of human IKKα (amino acids 718-732). This sequence is highly conserved in mouse IKKα (two amino acid substitutions).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1 % bovine serum albumin and 15 mM sodium azide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service