All Photos(1)

IR120PLUS

Sigma-Aldrich

Amberlite® Strongly Acidic Cation Exchanger

gel form

CAS Number:
MDL number:

form

gel form

contains

~45% moisture

feature

effective pH range 0 ‑ 14
operating temperature≤ 120 °C

matrix

8% cross-linked polystyrene

matrix active group

sulfonic acid

particle size

16-50 mesh (wet)

capacity

1.9 meq/mL
4.4 meq/g

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service