All Photos(1)
L1022
Supelco
Lamps, deuterium
For Beckman ACTA, 24, 25, 34, DU8
availability
not available in EU
Features and Benefits
A special optical window permits high intensity from 160 to 375 nm. Minimum 500 hr useful life.
Original-equipment quality and fit for spectrophotometer models listed
Compatibility
For UV spectrophotometers
Certificate of Analysis
Certificate of Origin