All Photos(1)

L2125

Sigma-Aldrich

L-Leucine tert-butyl ester hydrochloride

Linear Formula:
(CH3)2CHCH2CH(NH2)COOC(CH3)3 · HCl
CAS Number:
Molecular Weight:
223.74
Beilstein:
3697141
MDL number:
PubChem Substance ID:
NACRES:
NA.26

assay

≥98% (TLC)

Quality Level

form

powder

color

white

storage temp.

−20°C

SMILES string

Cl.CC(C)C[C@H](N)C(=O)OC(C)(C)C

InChI

1S/C10H21NO2.ClH/c1-7(2)6-8(11)9(12)13-10(3,4)5;/h7-8H,6,11H2,1-5H3;1H/t8-;/m0./s1

InChI key

RFUWRXIYTQGFGA-QRPNPIFTSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Naoatsu Shibata et al.
The Journal of organic chemistry, 75(16), 5707-5715 (2010-08-14)
Reversal of enantioselectivity in a Cu-catalyzed asymmetric conjugate addition reaction of dialkylzinc to cyclic enone with use of the same chiral ligand was successfully achieved. The reaction of 2-cyclohexen-1-one (30) with Et(2)Zn catalyzed by Cu(OTf)(2) in the presence of an
Cuie Wang et al.
Langmuir : the ACS journal of surfaces and colloids, 27(19), 12058-12068 (2011-08-23)
Infrared spectroscopy was applied to investigate the well-known EDC/NHS (N-ethyl-N'-(3-(dimethylamino)propyl)carbodiimide/N-hydroxysuccinimide) activation details of poly(acrylic acid) (PAA) and poly(methacrylic acid) (PMAA) brushes grafted on porous silicon. Succinimidyl ester (NHS-ester) is generally believed to be the dominant intermediate product, conveniently used to
Jan Pícha et al.
Amino acids, 39(5), 1265-1280 (2010-03-30)
In the present study, we describe in detail the synthesis of a relatively rare class of phosphorus compounds, α-carboxyphosphinopeptides. We prepared several norleucine-derived α-carboxyphosphinic pseudopeptides of the general formula Nle-Ψ[PO(OH)]-Gly. These compounds could have important applications as transition state-mimicking inhibitors
David Schachter et al.
American journal of physiology. Gastrointestinal and liver physiology, 306(11), G938-G946 (2014-04-05)
The leucine-to-glutamate (Leu→Glu) pathway, which metabolizes the carbon atoms of l-leucine to form l-glutamate, was studied by incubation of rat tissue segments with l-[U-(14)C]leucine and estimation of the [(14)C]glutamate formed. Metabolism of the leucine carbon chain occurs in most rat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service