All Photos(3)

M0823

Sigma-Aldrich

Anti-MKNK2 (402-415) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):
Anti-MNK2, Anti-MAP kinase-interacting serine/threonine kinase 2 isoform 2, Anti-GPRK7

biological source

rabbit

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~52 kDa

species reactivity

human

application(s)

western blot: 1:500-1:2,000

conjugate

unconjugated

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MKNK2(2872)

Immunogen

synthetic peptide corresponding to amino acids 402-415 of human MKNK2.

Application

Anti-MKNK2 (402-415) antibody produced in rabbit is suitable for western blot analysis at a dilution of 1:500-1:2,000.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Certificate of Origin

Dieter Dauwe et al.
Journal of the American Heart Association, 5(4), e002288-e002288 (2016-04-20)
Blood outgrowth endothelial cells (BOECs) mediate therapeutic neovascularization in experimental models, but outgrowth characteristics and functionality of BOECs from patients with ischemic cardiomyopathy (ICMP) are unknown. We compared outgrowth efficiency and in vitro and in vivo functionality of BOECs derived...
Sabine Fuchs et al.
Biomaterials, 30(7), 1329-1338 (2008-12-19)
For successful bone regeneration tissue engineered bone constructs combining both aspects, namely a high osteogenic potential and a rapid connection to the vascular network are needed. In this study we assessed the formation of pre-vascular structures by human outgrowth endothelial...
Ming Li et al.
Tissue engineering. Part C, Methods, 20(4), 328-339 (2013-09-04)
The repair and regeneration of large bone defects, including the formation of functional vasculature, represents a highly challenging task for tissue engineering and regenerative medicine. Recent studies have shown that vascularization and ossification can be stimulated by mild heat stress...
Peter Steinbacher et al.
PloS one, 10(4), e0123881-e0123881 (2015-04-18)
PGC-1α (peroxisome proliferator-activated receptor γ co-activator 1α) is an important regulator of mitochondrial biogenesis and a master regulator of enzymes involved in oxidative phosphorylation. Recent evidence demonstrated that the Gly482Ser single nucleotide polymorphism (SNP) in the PGC-1α gene affects insulin...
Marlen Kolbe et al.
Tissue engineering. Part C, Methods, 16(5), 877-886 (2009-11-07)
An effective isolation protocol for outgrowth endothelial cells (OEC) resulting in higher cell numbers and a reduced expansion time would facilitate the therapeutical application. In this study a standard protocol based on the isolation of mononuclear cells from adult peripheral...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service