All Photos(1)

M1501

Sigma-Aldrich

Methoxychlor

~95%, crystalline

Synonym(s):
2,2-Bis(4-methoxyphenyl)-1,1,1-trichloroethane, DMDT, 1,1,1-Trichloro-2,2-bis-(p-methoxyphenyl)ethane
Empirical Formula (Hill Notation):
C16H15Cl3O2
CAS Number:
Molecular Weight:
345.65
Beilstein:
2057367
EC Number:
MDL number:
PubChem Substance ID:

assay

~95%

form

crystalline

color

yellow

mp

86-88 °C (lit.)

SMILES string

COc1ccc(cc1)C(c2ccc(OC)cc2)C(Cl)(Cl)Cl

InChI

1S/C16H15Cl3O2/c1-20-13-7-3-11(4-8-13)15(16(17,18)19)12-5-9-14(21-2)10-6-12/h3-10,15H,1-2H3

InChI key

IAKOZHOLGAGEJT-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pesticide with estrogenic activity.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service