M8818

Sigma-Aldrich

Maize Polymorphic SSR Primer Set

Pricing and availability is not currently available.

storage temp.

−20°C

General description

The Maize Polymorphic SSR Primer Set is a cost-effective solution for SSR or SSLP mapping and screening projects that reduces or eliminates the need for custom oligos. The Set consists of 480 pairs of primers selected from the maize genome database. The selections cover the entire maize genome and target regions that have a high probability of polymorphism. This primer set is useful for screening new maize populations for genetic mapping, genotyping, marker-trait association analysis, and physical mapping of BAC clones, etc. All primers pass sequence and PCR validation tests. The primers are arrayed in five 96-well plates with a normalized concentration of 50 μM / 200 μl (1 nmol / well). Additional information on Sigma′s Maize SSR Primers can be found at www.sigma-aldrich.com/maizessr.

Packaging

  • Five 96-well plates containing 480 pairs of lyophilized primers (1 nmol per well)
  • Five additional 96-well plates for dilutions
  • 20 sheets of sealing films

Other Notes

The difference between the Maize SSR Primer Set (M4193) and the Maize Polymorphic SSR Primer Set (M8818) is the intention of the sets. The Maize SSR Primer Set is intended to provide relatively even coverage of the entire maize genome (average of 20 cM units of map distance between SSR markers). The Maize Polymorphic SSR Primer Set provides coverage of the entire maize genome, however, it is intended to target genome regions that have a high probability of polymorphism, with little regard to spacing between SSR markers.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.