MB1

Sigma-Aldrich

Methylene Blue hydrate

suitable for blood stain

Empirical Formula (Hill Notation):
C16H18ClN3S · xH2O
CAS Number:
Molecular Weight:
319.85 (anhydrous basis)
Beilstein/REAXYS Number:
3599847
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

(IR spectrum consistent with structure)

suitability

suitable for blood stain

SMILES string

O.Cl-.CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=N+(/C)C

InChI

1S/C16H18N3S.ClH.H2O/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;;/h5-10H,1-4H3;1H;1H2/q+1;;/p-1

InChI key

WQVSELLRAGBDLX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xn

Risk Statement

22

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Sadia Malik et al.
Microscopy research and technique, 83(12), 1594-1603 (2020-08-17)
This study aimed the optical imaging of malignant and normal skin tissues with multimodal wide-field fluorescence polarization imaging (WF) technique, by using methylene blue as fluorescence dye. We present optical imaging of skin tissues by different techniques, including reflectance, fluorescence...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.