All Photos(1)

N2757

Sigma-Aldrich

Nonanoic anhydride

~98%

Empirical Formula (Hill Notation):
C18H34O3
CAS Number:
Molecular Weight:
298.46
EC Number:
MDL number:
PubChem Substance ID:

assay

~98%

storage temp.

−20°C

SMILES string

CCCCCCCCC(=O)OC(=O)CCCCCCCC

InChI

1S/C18H34O3/c1-3-5-7-9-11-13-15-17(19)21-18(20)16-14-12-10-8-6-4-2/h3-16H2,1-2H3

InChI key

PBBAFLCORNAZCD-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service