N4770

Sigma-Aldrich

Nω-Nitro-D-arginine methyl ester hydrochloride

Synonym(s):
NG-Nitro-D-arginine methyl ester hydrochloride, D-NAME
Empirical Formula (Hill Notation):
C7H15N5O4 · HCl
CAS Number:
Molecular Weight:
269.69
MDL number:
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

storage temp.

−20°C

SMILES string

Cl.COC(=O)C(N)CCCNC(=N)NN(=O)=O

InChI

1S/C7H15N5O4.ClH/c1-16-6(13)5(8)3-2-4-10-7(9)11-12(14)15;/h5H,2-4,8H2,1H3,(H3,9,10,11);1H

InChI key

QBNXAGZYLSRPJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Inactive isomer of NAME

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Xi-Yong Yu et al.
PloS one, 6(1), e16239-e16239 (2011-02-02)
Diabetes has been regarded as an inflammatory condition which is associated with left ventricular diastolic dysfunction (LVDD). The purpose of this study was to examine the expression levels of macrophage migration inhibitory factor (MIF) and G protein-coupled receptor kinase 2...
Mauricio Sendeski et al.
Radiology, 251(3), 697-704 (2009-04-16)
To determine whether a type of contrast medium (CM), iodixanol, modifies outer medullary descending vasa recta (DVR) vasoreactivity and nitric oxide (NO) production in isolated microperfused DVR. Animal handling conformed to the Animal Care Committee Guidelines of all participating institutions....

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.