MilliporeSigma
All Photos(1)

N6033

Sigma-Aldrich

N-(NBD-Aminocaproyl)-L-threo-sphingosine

>98%

Synonym(s):
N-[6-[(7-Nitro-2-1,3-benzoxadiazol-4-yl)amino]caproyl]-L-threo-sphingosine, NBD-C6-L-Threo-sphingosine
Empirical Formula (Hill Notation):
C30H49N5O6
Molecular Weight:
575.74
MDL number:

assay

>98%

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](CO)NC(=O)CCCCCNc1ccc([N+]([O-])=O)c2nonc12

General description

Fluorescently labeled fatty acid sphingolipid membrane probe. NBD group excitation 460 nm, emission 534 nm.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service