MilliporeSigma
All Photos(1)

N6158

Sigma-Aldrich

N-(NBD-Aminocaproyl)sphingosine β-D-galactosyl

≥98%

Synonym(s):
C6-NBD Galactosyl cerebroside, N-[6-[(7-Nitro-2-1,3-benzoxadiazol-4-yl)amino]caproyl]-D-galactosyl-β1-1′-sphingosine
Empirical Formula (Hill Notation):
C36H59N5O11
Molecular Weight:
737.88
MDL number:
PubChem Substance ID:
NACRES:
NA.77

Quality Level

assay

≥98%

form

solid

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)NC(=O)CCCCCNc2ccc([N+]([O-])=O)c3nonc23

InChI

1S/C36H59N5O11/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-18-28(43)26(24-50-36-35(47)34(46)33(45)29(23-42)51-36)38-30(44)19-16-14-17-22-37-25-20-21-27(41(48)49)32-31(25)39-52-40-32/h15,18,20-21,26,28-29,33-37,42-43,45-47H,2-14,16-17,19,22-24H2,1H3,(H,38,44)/b18-15+/t26-,28+,29+,33-,34-,35+,36+/m0/s1

InChI key

ZYEQNDHFYIPTAY-LFUFHFQKSA-N

Related Categories

General description

Fluorescently labeled fatty acid sphingolipid membrane probe. NBD group excitation 460 nm, emission 534 nm.

Packaging

50 μg in serum bottle

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service