All Photos(1)

N7644

Sigma-Aldrich

(2-Nonen-1-yl)succinic anhydride

≥85% (titration)

Empirical Formula (Hill Notation):
C13H20O3
CAS Number:
Molecular Weight:
224.30
EC Number:
MDL number:
PubChem Substance ID:

assay

≥85% (titration)

density

1.03 g/mL at 20 °C (lit.)

SMILES string

CCCCCC\C=C\CC1CC(=O)OC1=O

InChI

1S/C13H20O3/c1-2-3-4-5-6-7-8-9-11-10-12(14)16-13(11)15/h7-8,11H,2-6,9-10H2,1H3/b8-7+

InChI key

CIMXGTDNUHWJCZ-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Special grade for electron microscopy: hardener for epoxy resins.

Certificate of Analysis

Certificate of Origin

A low-viscosity epoxy resin embedding medium for electron microscopy.
A R Spurr
Journal of ultrastructure research, 26(1), 31-43 (1969-01-01)
Designing multi-viscosity embedding media utilizing novel resin/anhydride/catalyst combinations.
J A Mascorro et al.
Microscopy research and technique, 20(1), 105-106 (1992-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service