NP40

Sigma-Aldrich

TERGITOL

Type NP-40

CAS Number:
MDL number:
Pricing and availability is not currently available.

description

non-ionic

type

Type NP-40

form

low-melting solid

Looking for similar products? Visit Product Comparison Guide

Other Notes

Polyglycol ether (nonionic) surfactants.
Do not confuse with Nonidet P-40, which is a liquid. Nonidet P-40 is no longer commercially available: Igepal CA-630 (CAS No. 9002-93-1) is offered as a replacement for the Nonidet P-40.

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xn,N

Risk Statement

20/22-41-51/53

Safety Statement

26-39-61

RIDADR

UN 3082 9 / PGIII

WGK Germany

WGK 2

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.