All Photos(1)

O3755

Sigma-Aldrich

Octadecylthioethyl 4-O-(4-O-[6-O-α-D-glucopyranosyl-α-D-glucopyranosyl]-α-D-glucopyranosyl)-β-D-glucopyranoside

≥95%

Synonym(s):
α-D-Glc-(1→6)-α-D-Glc-(1→4)-α-D-Glc-(1→4)-β-D-Glc-1→O-OTE
Empirical Formula (Hill Notation):
C44H82O21S
CAS Number:
Molecular Weight:
979.17
MDL number:
PubChem Substance ID:
NACRES:
NA.25

Quality Level

assay

≥95%

form

solid

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCSCCOC1OC(CO)C(OC2OC(CO)C(OC3OC(COC4OC(CO)C(O)C(O)C4O)C(O)C(O)C3O)C(O)C2O)C(O)C1O

InChI

1S/C44H82O21S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-66-20-18-58-41-37(56)33(52)39(26(22-46)61-41)65-44-38(57)34(53)40(27(23-47)62-44)64-43-36(55)32(51)30(49)28(63-43)24-59-42-35(54)31(50)29(48)25(21-45)60-42/h25-57H,2-24H2,1H3

InChI key

LCVLXPYSKDKYEK-UHFFFAOYSA-N

Related Categories

Certificate of Analysis

Certificate of Origin

P Hallgren et al.
The Journal of biological chemistry, 252(3), 1034-1040 (1977-02-10)
The excretion rates of different oligosaccharides in the urine of two individuals, a blood group O, nonsecretor woman, and a blood group A, secretor woman, were studied at different times during pregnancy and lactation. Quantitative determinations of di-, tri-, and
M Kuriyama et al.
Journal of biochemistry, 98(4), 1041-1047 (1985-10-01)
Neutral oligosaccharides were isolated from urine of an adult patient with glycogen storage disease type II, a deficiency of lysosomal acid alpha-glucosidase, by chromatography on columns of activated charcoal, Dowex 50 X 2 and Dowex 1 X 2. Total neutral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service