MilliporeSigma
All Photos(1)

O9632

Sigma-Aldrich

d(pT)12 ammonium salt

~90%, lyophilized powder

assay

~90%

form

lyophilized powder

storage temp.

−20°C

Preparation Note

Prepared by automated DNA synthesis.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service