P0905

Sigma-Aldrich

Phosphatase, Alkaline Type XXX-T from calf intestinal mucosa

buffered aqueous solution, 2,000-4,000 DEA units/mg protein, 30-90 units/mg protein (modified Warburg-Christian)

Pricing and availability is not currently available.

form

buffered aqueous solution

specific activity

2,000-4,000 DEA units/mg protein
30-90 units/mg protein (modified Warburg-Christian)

storage temp.

2-8°C

Packaging

Package size based on DEA units

Unit Definition

One DEA unit will hydrolyze 1 μmole of 4-nitrophenyl phosphate per minute at pH 9.8 at 37°C.2

Physical form

Solution in 3.0 M NaCl containing 1 mM MgCl2, 0.1 mM ZnCl2 and 30 mM triethanolamine, pH 7.6

Preparation Note

Affinity purified. Prepared from P 4028, Type XXX

Analysis Note

Protein determined by biuret unless otherwise indicated.

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.