All Photos(2)

P1231

Sigma-Aldrich

Anti-P2Y4 Purinergic Receptor antibody produced in rabbit

affinity isolated antibody, lyophilized powder

MDL number:

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

application(s)

western blot (chemiluminescent): 1:300

conjugate

unconjugated

UniProt accession no.

storage temp.

−20°C

Gene Information

Immunogen

synthetic peptide corresponding to amino acids 337-350 of rat P2Y4.

Physical form

Lyophilized at 0.3 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose, and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Certificate of Origin

Y D Bogdanov et al.
British journal of pharmacology, 124(3), 428-430 (1998-07-01)
An intronless open reading frame encoding a protein (361aa in length) was isolated from a rat genomic library probed with a DNA fragment from rat heart. This protein showed 83% sequence identity with the human P2Y4 (hP2Y4) receptor and represents...
M M Ward et al.
Neuroscience, 155(4), 1262-1274 (2008-07-16)
Extracellular ATP acts as a neurotransmitter in the retina, via the activation of ionotropic P2X receptors and metabotropic P2Y receptors. The expression of various P2X and P2Y receptor subtypes has been demonstrated in the retina, but the localization of P2Y...
T E Webb et al.
Journal of neurochemistry, 71(4), 1348-1357 (1998-09-29)
Degenerate PCR was used to amplify DNAs encoding members of the P2Y receptor family from rat brain RNA. A full-length sequence obtained for one novel clone (R5) contained an intronless open reading frame that encoded a polypeptide of 361 amino...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service