MilliporeSigma
All Photos(1)

P1551

Sigma-Aldrich

16-Hydroxyphorbol 12-palmitate 13-acetate

Synonym(s):
16-OH-Phorbol 12-palmitate-13-acetate, 12-O-Hexadecanoyl-16-hydroxyphorbol-13-acetate, HHPA
Empirical Formula (Hill Notation):
C38H60O9
CAS Number:
Molecular Weight:
660.88
Beilstein:
2799897
MDL number:

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)[C@@H](C=C(CO)C[C@@]3(O)[C@H]2C=C(C)C3=O)[C@@H]4[C@@](C)(CO)[C@]14OC(C)=O

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service