P2362

Greiner PCR 96 well plates, polycarbonate

Type 2 for Hybaid, 96 well

Synonym(s):
96 multiwell plates, 96 well microtiter plates, 96 well plates, 96 well microplates

material

clear polycarbonate, type 2

sterility

non-sterile

packaging

case of 50

manufacturer/tradename

Greiner 651550

application(s)

PCR: suitable

size

96 well

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

ThermoQuick 96W PCR Microplate, well working volume 200 μL.
PCR Plates have a standard microplate footprint and are compatible with thermocyclers of most major brands. DNAse and RNAse free and non-pyrogenic by LAL method to <0.06 EU/ml.  

Other Notes

96 well microplate, without lid, no skirt alphanumeric well coding, length: 126.50 mm; width: 86; mm thickness 635 μm.

Suitability

Use for polymerase chain reaction; suitable for Hybaid thermocycler, Optimal and rapid heat transmission; Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service