All Photos(1)

P3610

Sigma-Aldrich

Anti-P2X3 Purinergic Receptor antibody produced in rabbit

100 μg/mL, affinity isolated antibody, buffered aqueous solution

MDL number:

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

concentration

100 μg/mL

application(s)

immunohistochemistry: suitable
indirect immunofluorescence: suitable
western blot: suitable

conjugate

unconjugated

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... P2RX3(5024)
mouse ... P2rx3(228139)
rat ... P2rx3(81739)

Immunogen

highly purified peptide corresponding to amino acids 383-397 from the rat P2X3 receptor conjugated to KLH.

Physical form

Solution in 0.05 M sodium phosphate buffer containing 0.1% sodium azide and 0.2% gelatin.

Certificate of Analysis

Certificate of Origin

R A North et al.
Current opinion in neurobiology, 7(3), 346-357 (1997-06-01)
Adenosine 5'-triphosphate (ATP) and/or related nucleotides act at both ionotropic (P2X) and metabotropic (P2Y) receptors. P2X receptor subunits (P2X1-P2X7) form ligand-gated cation channels, as homomultimers or heteromultimers. Recent work indicates that P2X3 subunits participate in channels expressed by nociceptive sensory...
Receptors for purines and pyrimidines.
V Ralevic et al.
Pharmacological reviews, 50(3), 413-492 (1998-10-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service