MilliporeSigma
All Photos(1)

P4275

Sigma-Aldrich

N-(10-[1-Pyrene]decanoyl)sphingomyelin

~95%

assay

~95%

storage temp.

−20°C

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service