P5401

Sigma-Aldrich

Ethyl petroselaidate

Synonym(s):
Petroselaidic acid ethyl ester
Empirical Formula (Hill Notation):
C20H38O2
CAS Number:
Molecular Weight:
310.51
MDL number:
PubChem Substance ID:

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCC\C=C\CCCCC(=O)OCC

InChI

1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h14-15H,3-13,16-19H2,1-2H3/b15-14+

InChI key

IKUZCNYUCLNCKN-CCEZHUSRSA-N

Related Categories

storage_class_code

12 - Non Combustible Liquids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service