P5526

Sigma-Aldrich

Methyl petroselaidate

≥99%

Synonym(s):
Methyl trans-6-octadecenoate, Petroselaidic acid methyl ester
Empirical Formula (Hill Notation):
C19H36O2
CAS Number:
Molecular Weight:
296.49
EC Number:
MDL number:

assay

≥99%

storage temp.

−20°C

SMILES string

CCCCCCCCCCC\C=C\CCCCC(=O)OC

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service