Documents

P7977

Sigma-Aldrich

Proli NONOate

Synonym(s):
Disodium 1-(2-carboxylato)pyrrolidin-1-yldiazen-1-ium-1,2-diolate methanol
Linear Formula:
C5H7N3O4Na2
Molecular Weight:
219.11
MDL number:
Pricing and availability is not currently available.

storage temp.

−20°C

Application

Nitric oxide donor useful for reliable generation of nitric oxide in vitro and in vivo.
Certificate of Analysis
Certificate of Origin
Kiefer, L.K., et al.
Methods in Enzymology, 298, 281-281 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.