Documents

P8402

Sigma-Aldrich

Protease from Bacillus sp.

lyophilized powder, 200-300 units/mg protein (Lowry)

EC Number:

form

lyophilized powder

specific activity

200-300 units/mg protein (Lowry)

storage temp.

2-8°C

Unit Definition

One unit will hydrolyze 1.0 μmole of Nα-p-tosyl-L-arginine methyl ester per min at pH 8.0 at 25°C.

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.