MilliporeSigma
All Photos(1)

P8851

Millipore

Petragnani medium base

Application

Prepared for the isolation and cultivation of Mycobacterium.

Components

Ingredients (g/L)
Casein enzymic hydrolysate, 5.10
L-Asparagine, 5.10
Potato flour, 36.40
Skim milk, 100.00
Malachite green, 1.20

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service