All Photos(1)

Q2628

Sigma-Aldrich

QAE Cellulose Fast Flow (QA-92)

fibers

Synonym(s):
Diethyl-(2-hydroxypropyl)aminoethyl cellulose, QAE cellulose
MDL number:

form

fibers

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service