R1501

Sigma-Aldrich

Ricinelaidic acid sodium salt

~99%

Synonym(s):
(R)-12-Hydroxy-9(E)-octadecenoic acid
Linear Formula:
C18H33O3Na
CAS Number:
Molecular Weight:
320.44
MDL number:
PubChem Substance ID:

assay

~99%

form

solid

functional group

carboxylic acid

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na].CCCCCCC(O)C\C=C\CCCCCCCC(O)=O

InChI

1S/C18H34O3.Na.H/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21;;/h9,12,17,19H,2-8,10-11,13-16H2,1H3,(H,20,21);;/b12-9+;;

InChI key

CETAEINSXPFLIW-ANOGCNOSSA-N

Packaging

Sealed ampule.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service