R3750

Sigma-Aldrich

Retinyl palmitate

Type VII, matrix dispersion, ~250,000 USP units/g

Synonym(s):
Vitamin A palmitate, all−trans−Retinol palmitate
Empirical Formula (Hill Notation):
C36H60O2
CAS Number:
Molecular Weight:
524.86
Beilstein/REAXYS Number:
1917366
EC Number:
MDL number:
Pricing and availability is not currently available.

type

Type VII

form

matrix dispersion

specific activity

~250,000 USP units/g

contains

butylated hydroxytoluene as antioxidant
butylated hydroxyanisole

storage temp.

2-8°C

SMILES string

CC1=C(/C=C/C(C)=C/C=C/C(C)=C/COC(CCCCCCCCCCCCCCC)=O)C(C)(C)CCC1

InChI

1S/C36H60O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-25-35(37)38-30-28-32(3)23-20-22-31(2)26-27-34-33(4)24-21-29-36(34,5)6/h20,22-23,26-28H,7-19,21,24-25,29-30H2,1-6H3/b23-20+,27-26+,31-22+,32-28+

InChI key

VYGQUTWHTHXGQB-FFHKNEKCSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Review: Vitamin A metabolism.

Physical form

Dispersion in acacia-starch matrix. Water dispersible

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.