R492-05A

Sigma-Aldrich

Rat Marrow Stromal Cells: RMSC, adult

NACRES:
NA.81
Pricing and availability is not currently available.

biological source

rat bone marrow (Rat bone marrow)

Quality Level

packaging

pkg of 500,000 cells

growth mode

Adherent

morphology

stromal

application(s)

cell culture | mammalian: suitable

relevant disease(s)

tuberculosis

shipped in

dry ice

storage temp.

−196°C

Related Categories

General description

Bone marrow is the major blood creating organ, but in addition to supporting hematopoietic growth and differentiation, marrow stromal cells can be induced to produce cells of other connective tissues, such as bone, cartilage, and fat, as well as cells from neuroectodermal (neurons) and endodermal (hepatocytes) lineages. The potential of HMSCs to maintain multipotency and proliferate extensively in vitro provides new avenues for cell-based therapy in the restoration of damaged or diseased tissue.RMSC from Cell Applications, Inc. have been used to demonstrate that TGF-β stimulates production of MCP-1 by vascular smooth muscle cells, which attracts bone marrow stromal cells (Zhang, 2009); and show that their migration can be stimulated by VPA and lithium through HDAC-CXCR4 and GSK-3β-MMP-9, respectively (Tsai, 2010); to investigate the therapeutic potential of marrow stromal stem cells depending on the extracellular matrix properties (Gershlak, 2013; Sullivan, 2014); and demonstrate that combination of low-level laser therapy and transplantation of marrow stromal stem cells results in greater functional recovery after nerve crush injury (Yang, 2013). Finally, HMSC were used to develop methods to direct stem cell differentiation by altering physical topography of the substrate (Brammer, 2011), and to design surface materials to control cell-adhesion properties (Sommani, 2010; Tsuji, 2011)

Cell Line Origin

Bone Marrow

Application

Blood production, hematopoietic growth and differentiation, pathways of pluripotent cell differentiation into bone, cartilage and fat, connective tissue production,supportive structure, cell production, cell proliferation, cell based therapy, tissue restoration and repair, differentiation, effects of culture conditinos, HLA expression, cytokine production, immunomodulation, clinical trials.

Components

Rat Marrow Stromal Cell Basal Medium that contains 10% FBS and 10% DMSO

Quality

Each lot was tested for proper morphology, Population Doublings, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.

Preparation Note

  • 2nd passage, >500,000 cells in Rat Marrow Stromal Cell Basal Medium that contains 10% FBS and 10% DMSO
  • Can be cultured at least 10 population doublings

Subculture Routine

Please refer to the RMSC Culture Protocol.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.