R7135

Sigma-Aldrich

Sph I from Streptomyces phaeochromogenes

Restriction Enzyme

CAS Number:
Enzyme Commission number:
MDL number:
Pricing and availability is not currently available.

grade

for molecular biology

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Specificity

Recognition sequence: 5′-GCATG/C-3′
Cutting results: A 2-10-fold Sph I overdigestion of 1 μg λ DNA substrate results in 100% cutting.
Heat inactivation: Inactivated at 65 °C for 20 minutes.

Other Notes

Supplied with 10x Restriction Enzyme Buffer SM (B3158).

Physical form

Solution in 20 mM Tris-HCl , pH 8.2, 1 mM EDTA, 350 mM NaCl, 7 mM 2-mercaptoethanol, 0.1 mM PMSF, 50% glycerol (v/v), 0.2% Triton X-100 (v/v), at 4 °C

RIDADR

NONH for all modes of transport

WGK Germany

1

Certificate of Analysis
Certificate of Origin
Characterization of a site-specific restriction endonuclease Sph l from Streptomyces phaeochromogenes.
Fuchs, L.Y., et al.
Gene, 10, 391-391 (1980)
C Kessler et al.
Gene, 92(1-2), 1-248 (1990-08-16)
The properties and sources of all known class-I, class-II and class-III restriction endonucleases (ENases) and DNA modification methyltransferases (MTases) are listed and newly subclassified according to their sequence specificity. In addition, the enzymes are distinguished in a novel manner according...
Jaroslav Jelinek et al.
Epigenetics, 7(12), 1368-1378 (2012-10-19)
Genome wide analysis of DNA methylation provides important information in a variety of diseases, including cancer. Here, we describe a simple method, Digital Restriction Enzyme Analysis of Methylation (DREAM), based on next generation sequencing analysis of methylation-specific signatures created by...
Saqib H Ansari et al.
Journal of pediatric hematology/oncology, 35(4), e153-e156 (2013-02-08)
β-thalassemia is characterized by impaired β-chain synthesis leading to ineffective erythropoiesis, severe anemia, and a need for blood transfusion. Presence of Xmn I polymorphism (-158 C-T nucleotide change) in γ-globin gene is associated with a higher fetal hemoglobin and a...
Amal M El-Shehaby et al.
Scandinavian journal of clinical and laboratory investigation, 73(1), 75-81 (2012-12-04)
Left ventricular hypertrophy (LVH) is a common manifestation of cardiovascular disease and has an important prognostic value in patients with end-stage renal disease (ESRD). Vitamin D receptor (VDR) has been intensively investigated, and one of these (BsmI) already has been...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.