Documents

R9376

Sigma-Aldrich

AMV-Reverse Transcriptase from avian myeloblastosis virus

Synonym(s):
RNA-directed DNA polymerase, Revertase
CAS Number:
Enzyme Commission number:
MDL number:
Pricing and availability is not currently available.

form

solution

concentration

10,000-100,000 units/mL

shipped in

dry ice

storage temp.

−70°C

Application

AMV reverse transcriptase synthesizes DNA complementary (cDNA) to RNA templates. A DNA primer complementary to the RNA template and a divalent cation, either Mg or Mn, are required for initiation of transcription. This enzyme is commonly used to make cDNAs from mRNA for eventual cloning or for use as probes.

Unit Definition

One unit incorporates 1 nmol of dTTP into TCA precipitable material in 10 min at 37 °C using polyadenylic acid as template and oligo(dT)12-18 as a primer.

Legal Information

Purchase of this product is accompanied by a limited license for use in the Polymerase Chain Reaction (PCR) process for research purposes only and in conjunction with a thermal cycler whose use in the automated performance of the PCR process is covered by an up-front license fee, either by payment to Applied Biosystems or as purchased, i.e., and authorized thermal cycler.

WGK Germany

1

Christopher M Bailey et al.
Journal of medicinal chemistry, 56(10), 3959-3968 (2013-05-11)
Human immunodeficiency virus type 1 reverse transcriptase (HIV-1 RT) is a major target for currently approved anti-HIV drugs. These drugs are divided into two classes: nucleoside and non-nucleoside reverse transcriptase inhibitors (NRTIs and NNRTIs). This study illustrates the synthesis and...
Xiu-Juan Jiao et al.
Zhonghua shi yan he lin chuang bing du xue za zhi = Zhonghua shiyan he linchuang bingduxue zazhi = Chinese journal of experimental and clinical virology, 26(6), 453-455 (2013-05-01)
To characterize genotypic resistance within HBV RT region in chronic hepatitis B (CHB) patients with nucleos(t)ide analogue (NA) treatment. Serum samples of 229 CHB patients with NA treatment were obtained. Full-length HBV RT sequences were amplified, sequenced and analyzed, on...
TERT hypermethylation: biomarker in paediatric brain tumours.
Scott J Diede
The Lancet. Oncology, 14(6), 447-448 (2013-04-20)
Jiansen Jiang et al.
Nature, 496(7444), 187-192 (2013-04-05)
Telomerase adds telomeric repeats to chromosome ends using an internal RNA template and a specialized telomerase reverse transcriptase (TERT), thereby maintaining genome integrity. Little is known about the physical relationships among protein and RNA subunits within a biologically functional holoenzyme....
Ausubel, F., et al.
Short Protocols in Molecular Biology, 14-14 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.