All Photos(1)

S0541

Sigma-Aldrich

Smilagenin

≥98%

Synonym(s):
16,22:22,26-Bis(epoxy)cholestan-3β-ol, (25R)-Spirostan-3β-ol
Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
Beilstein:
91753
EC Number:
MDL number:

assay

≥98%

SMILES string

[H][C@]12CCC3C(CC[C@@]4(C)C3C[C@@]5([H])O[C@]6(CC[C@@H](C)CO6)[C@@H](C)[C@]45[H])[C@@]1(C)CC[C@H](O)C2

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service