MilliporeSigma
All Photos(1)

S2546

Sigma-Aldrich

[D-Trp7,9,10]-Substance P

≥97% (HPLC)

Synonym(s):
Arg-Pro-Lys-Pro-Gln-Gln-D-Trp-Phe-D-Trp-D-Trp-Met-NH2
Empirical Formula (Hill Notation):
C79H105N21O13S
CAS Number:
Molecular Weight:
1588.88
MDL number:

assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@@H](Cc1c[nH]c2ccccc12)NC(=O)[C@@H](Cc3c[nH]c4ccccc34)NC(=O)[C@H](Cc5ccccc5)NC(=O)[C@H](Cc6c[nH]c7ccccc67)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]8CCCN8C(=O)[C@H](CCCCN)NC(=O)[C@@H]9CCCN9C(=O)[C@@H](N)CCCNC(N)=N)C(N)=O

Biochem/physiol Actions

Substance P antagonist

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service